KEY465195715 Key Organics / BIONET

(2Z)-2-(4-chlorobenzenesulfonyl)-1-(4-chlorophenyl)-3-(dimethylamino)prop-2-en-1-one

  • MDL number MFCD00138912
  • Cas Number : 1164563-97-6
  • Molecular Weight : 384.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74056

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4D-015_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart