KEY465195717 Key Organics / BIONET

4-(4-chlorophenyl)-5,7-dihydroxy-3,4-dihydro-2H-1-benzopyran-2-one

  • MDL number MFCD00138915
  • Cas Number : 338417-72-4
  • Molecular Weight : 290.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74062

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4D-025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart