KEY465195722 Key Organics / BIONET

(3Z)-3-[(4-chlorophenyl)sulfanyl]-4-(dimethylamino)but-3-en-2-one

  • MDL number MFCD00138921
  • Cas Number : 76511-79-0
  • Molecular Weight : 255.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74073

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4D-051_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart