KEY465195723 Key Organics / BIONET

(2Z)-1-(4-chlorophenyl)-3-(dimethylamino)-2-(4-methylbenzenesulfonyl)prop-2-en-1-one

  • MDL number MFCD00138924
  • Cas Number : 338417-78-0
  • Molecular Weight : 363.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74077

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4D-060_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart