KEY465195735 Key Organics / BIONET

N-methyl-2-(pyridin-2-yl)-6-(trifluoromethyl)pyrimidin-4-amine

  • MDL number MFCD00139797
  • Cas Number : 338418-10-3
  • Molecular Weight : 254.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74124

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4E-310S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart