KEY465195738 Key Organics / BIONET

4-(prop-2-yn-1-ylsulfanyl)-2-(pyridin-2-yl)-6-(trifluoromethyl)pyrimidine

  • MDL number MFCD00139809
  • Cas Number : 338418-25-0
  • Molecular Weight : 295.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74140

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4E-333S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart