KEY465195739 Key Organics / BIONET

N-(7-hydroxy-2-oxo-2H-chromen-3-yl)benzamide

  • MDL number MFCD00140059
  • Cas Number : 126940-57-6
  • Molecular Weight : 281.267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74149

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4E-904_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart