KEY465195742 Key Organics / BIONET

methyl 6-methyl-2-oxo-3-[3-(trifluoromethyl)benzamido]-2H-pyran-5-carboxylate

  • MDL number MFCD00139818
  • Cas Number : 338418-34-1
  • Molecular Weight : 355.269

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74154

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4E-910_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart