KEY465195746 Key Organics / BIONET

methyl (3E)-3-{[(2,4-dichlorophenyl)methoxy]imino}-2-(phenylformamido)propanoate

  • MDL number MFCD00140979
  • Cas Number : 1214872-97-5
  • Molecular Weight : 395.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74162

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4E-921_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart