KEY465195747 Key Organics / BIONET

2-[(1Z)-1-cyano-2-phenyleth-1-en-1-yl]-5,7-dimethylpyrazolo[1,5-a]pyrimidine-3-carbonitrile

  • MDL number MFCD00141786
  • Cas Number : 1164556-51-7
  • Molecular Weight : 299.337

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link http://www.keyorganics.net/bionet/catalog/product/view/id/74168

Safety Information

signalword Warning
msds https://www.keyorganics.net/bionet/media/catalog/product/sds/4F-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart