KEY465195748 Key Organics / BIONET

2-[(1Z)-1-cyano-2-(pyridin-2-yl)eth-1-en-1-yl]-5,7-dimethylpyrazolo[1,5-a]pyrimidine-3-carbonitrile

  • MDL number MFCD00141787
  • Cas Number : 338418-44-3
  • Molecular Weight : 300.325

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74169

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-004_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart