KEY465195751 Key Organics / BIONET

5-[1-amino-2-(2,4-dichlorophenoxy)ethylidene]-2,2-dimethyl-1,3-dioxane-4,6-dione

  • MDL number MFCD00141614
  • Cas Number : 338418-52-3
  • Molecular Weight : 346.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74180

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-020_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart