KEY465195755 Key Organics / BIONET

2-[(4-cyano-5-methyl-1,2-thiazol-3-yl)sulfanyl]-N'-[2-(4-fluorophenoxy)ethanimidoyl]acetohydrazide

  • MDL number MFCD00214751
  • Cas Number : 338418-60-3
  • Molecular Weight : 379.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74188

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-029_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart