KEY465195757 Key Organics / BIONET

5-methyl-3-({[5-(phenoxymethyl)-1H-1,2,4-triazol-3-yl]methyl}sulfanyl)-1,2-thiazole-4-carbonitrile

  • MDL number MFCD00214754
  • Cas Number : 338418-70-5
  • Molecular Weight : 343.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74195

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4F-040_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart