KEY465195765 Key Organics / BIONET

3-(4-chlorophenyl)-1-[(1Z)-1-(2-phenylhydrazin-1-yl)-2-(1H-1,2,4-triazol-1-yl)ethylidene]urea

  • MDL number MFCD00202290
  • Cas Number : 338418-84-1
  • Molecular Weight : 369.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74213

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-060_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart