KEY465195770 Key Organics / BIONET

[1-(4-chlorophenyl)-1H-1,2,3-triazol-4-yl]methyl 3-(trifluoromethyl)benzoate

  • MDL number MFCD00140950
  • Cas Number : 338419-27-5
  • Molecular Weight : 381.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74286

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-381S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart