KEY465195771 Key Organics / BIONET

[1-(4-chlorophenyl)-1H-1,2,3-triazol-4-yl]methyl 4-chlorobutanoate

  • MDL number MFCD00140952
  • Cas Number : 338419-29-7
  • Molecular Weight : 314.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74289

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-384S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart