KEY465195774 Key Organics / BIONET

methyl 2-{[4-cyano-1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]sulfanyl}benzoate

  • MDL number MFCD00140982
  • Cas Number : 318239-35-9
  • Molecular Weight : 341.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74295

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-392S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart