KEY465195775 Key Organics / BIONET

methyl 3-{[3-(trifluoromethyl)phenyl]methoxy}thieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD00140985
  • Cas Number : 338419-35-5
  • Molecular Weight : 367.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74301

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-403S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart