KEY465195782 Key Organics / BIONET

(1-phenyl-1H-1,2,3-triazol-4-yl)methyl 2,4-dichlorobenzoate

  • MDL number MFCD00141073
  • Cas Number : 141581-65-9
  • Molecular Weight : 348.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74315

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-418S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart