KEY465195786 Key Organics / BIONET

5-(4-chlorobenzenesulfonyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD00140995
  • Cas Number : 318239-39-3
  • Molecular Weight : 349.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74325

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4F-428S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart