KEY465195792 Key Organics / BIONET

ethyl 5-(4-chlorophenyl)-2-(phenylhydrazido)-1H-pyrrole-3-carboxylate

  • MDL number MFCD00171814
  • Cas Number : 338419-67-3
  • Molecular Weight : 383.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74360

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-941_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart