KEY465195796 Key Organics / BIONET

3-[(E)-2-[(4-chlorophenyl)methylidene]hydrazin-1-yl]-1,2-dihydroquinoxalin-2-one

  • MDL number MFCD00139695
  • Cas Number : 129227-26-5
  • Molecular Weight : 298.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74367

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-948_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart