KEY465195797 Key Organics / BIONET

3-[(E)-2-[(2Z)-3-phenylprop-2-en-1-ylidene]hydrazin-1-yl]-1,2-dihydroquinoxalin-2-one

  • MDL number MFCD00139696
  • Cas Number : 338419-76-4
  • Molecular Weight : 290.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74368

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-949_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart