KEY465195800 Key Organics / BIONET

ethyl 5-(4-chlorophenyl)-2-[(4-chlorophenyl)amino]-1H-pyrrole-3-carboxylate

  • MDL number MFCD00171815
  • Cas Number : 338419-78-6
  • Molecular Weight : 375.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74373

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4F-954_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart