KEY465195803 Key Organics / BIONET

ethyl (2E)-3-amino-3-[(pyridin-4-yl)formohydrazido]prop-2-enoate

  • MDL number MFCD00243956
  • Cas Number : 146255-38-1
  • Molecular Weight : 250.258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74378

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-963_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart