KEY465195809 Key Organics / BIONET

4-(5-bromo-6-methoxynaphthalen-2-yl)-5-methylpyrimidin-2-amine

  • MDL number MFCD00173456
  • Cas Number : 338420-04-5
  • Molecular Weight : 344.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74393

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4G-014_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart