KEY465195811 Key Organics / BIONET

3,6-bis(3,5-dimethyl-1H-pyrazol-1-yl)-1,2-dihydro-1,2,4,5-tetrazine

  • MDL number MFCD00173231
  • Cas Number : 30169-21-2
  • Molecular Weight : 272.316

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74397

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4G-018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart