KEY465195816 Key Organics / BIONET

5-(2,4-dichlorophenyl)-12-(trifluoromethyl)-2,3,4,7,10-pentaazatricyclo[7.4.0.0^{2,6}]trideca-1(9),3,5,7,10,12-hexaen-8-amine

  • MDL number MFCD00173467
  • Cas Number : 338420-39-6
  • Molecular Weight : 399.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74423

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4G-055_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart