KEY465195817 Key Organics / BIONET

5-(4-chlorophenyl)-12-(trifluoromethyl)-2,3,4,7,10-pentaazatricyclo[7.4.0.0^{2,6}]trideca-1(9),3,5,7,10,12-hexaen-8-amine

  • MDL number MFCD00173468
  • Cas Number : 338420-41-0
  • Molecular Weight : 364.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74424

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4G-056_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart