KEY465195826 Key Organics / BIONET

5-(2-phenyl-1,3-thiazol-4-yl)-4-(prop-2-en-1-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00171868
  • Cas Number : 338420-11-4
  • Molecular Weight : 300.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74517

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4G-417S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart