KEY465195827 Key Organics / BIONET

prop-2-en-1-yl 6-chloro-2H-chromene-3-carboxylate

  • MDL number MFCD00202039
  • Cas Number : 338420-13-6
  • Molecular Weight : 250.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74520

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4G-421S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart