KEY465195828 Key Organics / BIONET

3,3-dimethyl 5-(4-chlorophenyl)-4,6-dioxo-hexahydro-2H-pyrrolo[3,4-d][1,2]oxazole-3,3-dicarboxylate

  • MDL number MFCD00663822
  • Cas Number : 338420-26-1
  • Molecular Weight : 368.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74533

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4G-911_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart