KEY465195829 Key Organics / BIONET

(1R,2R,6S)-4-(4-chlorophenyl)-7-oxa-4,8-diazatricyclo[6.4.0.0^{2,6}]dodecane-3,5-dione

  • MDL number MFCD00244768
  • Cas Number : 318239-72-4
  • Molecular Weight : 306.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74542

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4G-923_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart