KEY465195834 Key Organics / BIONET

(2E)-7-fluoro-3-phenyl-2-({[3-(trifluoromethyl)phenyl]amino}methylidene)-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD00232248
  • Cas Number : 338420-85-2
  • Molecular Weight : 397.373

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74589

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart