KEY465195835 Key Organics / BIONET

(2E)-7-fluoro-2-{[(2-nitrophenyl)amino]methylidene}-3-phenyl-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD00232249
  • Cas Number : 338420-86-3
  • Molecular Weight : 374.371

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74590

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-026_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart