KEY465195836 Key Organics / BIONET

methyl 1-methyl-2-{[(4-methylphenyl)methyl]sulfanyl}-1H-imidazole-5-carboxylate

  • MDL number MFCD00244348
  • Cas Number : 338420-88-5
  • Molecular Weight : 276.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74592

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-028_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart