KEY465195839 Key Organics / BIONET

potassium {5-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]pyrimidin-2-yl}sulfanide

  • MDL number MFCD02571807
  • Cas Number : 1007069-90-0
  • Molecular Weight : 329.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74596

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-033_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart