KEY465195841 Key Organics / BIONET

2-[(3-nitropyridin-2-yl)sulfanyl]-4-[(E)-2-phenylethenyl]pyrimidine

  • MDL number MFCD00244249
  • Cas Number : 338420-95-4
  • Molecular Weight : 336.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74601

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-039_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart