KEY465195846 Key Organics / BIONET

2-({[(2-methylphenyl)carbamoyl]methyl}sulfanyl)acetic acid

  • MDL number MFCD02594712
  • Cas Number : 436087-38-6
  • Molecular Weight : 239.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142 - 144
Purity 90
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74636

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-343S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart