KEY465195848 Key Organics / BIONET

3-cyano-4,6-dimethylpyridin-2-yl 2,4,6-trichlorobenzoate

  • MDL number MFCD00202385
  • Cas Number : 338421-29-7
  • Molecular Weight : 355.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74650

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-361S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart