KEY465195851 Key Organics / BIONET

2-({[(2,6-dichlorophenyl)carbamoyl]methyl}sulfanyl)acetic acid

  • MDL number MFCD00202388
  • Cas Number : 338421-32-2
  • Molecular Weight : 294.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74654

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-365S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart