KEY465195858 Key Organics / BIONET

methyl 2-({5-[(4-chlorobenzenesulfonamido)methyl]-4-methyl-4H-1,2,4-triazol-3-yl}sulfanyl)acetate

  • MDL number MFCD00180813
  • Cas Number : 338421-53-7
  • Molecular Weight : 390.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74679

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-406S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart