KEY465195864 Key Organics / BIONET

1-[(4-chlorophenyl)sulfanyl]-3-[4-(4-fluorophenyl)piperazin-1-yl]propan-2-ol

  • MDL number MFCD00244562
  • Cas Number : 338421-73-1
  • Molecular Weight : 380.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74704

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-443S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart