KEY465195865 Key Organics / BIONET

N-{[5-(butan-2-ylsulfanyl)-4-methyl-4H-1,2,4-triazol-3-yl]methyl}-4-fluorobenzene-1-sulfonamide

  • MDL number MFCD00180823
  • Cas Number : 338421-75-3
  • Molecular Weight : 358.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74706

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-445S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart