KEY465195866 Key Organics / BIONET

4-chloro-N-{[4-(prop-2-en-1-yl)-5-sulfanyl-4H-1,2,4-triazol-3-yl]methyl}benzene-1-sulfonamide

  • MDL number MFCD00180829
  • Cas Number : 338421-81-1
  • Molecular Weight : 344.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74712

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-453S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart