KEY465195875 Key Organics / BIONET

4-chloro-N-{[5-(methylsulfanyl)-4-(prop-2-en-1-yl)-4H-1,2,4-triazol-3-yl]methyl}benzene-1-sulfonamide

  • MDL number MFCD00202126
  • Cas Number : 338422-19-8
  • Molecular Weight : 358.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74753

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-501S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart