KEY465195882 Key Organics / BIONET

{1-[2-(3,4-dimethoxyphenyl)ethyl]-2-sulfanyl-1H-imidazol-5-yl}methanol

  • MDL number MFCD00215155
  • Cas Number : 338422-51-8
  • Molecular Weight : 294.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74791

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-931_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart