KEY465195884 Key Organics / BIONET

methyl 1-[4-cyano-5-(methylamino)-1,2-thiazol-3-yl]piperidine-4-carboxylate

  • MDL number MFCD00243962
  • Cas Number : 338422-53-0
  • Molecular Weight : 280.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74794

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-935_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart