KEY465195887 Key Organics / BIONET

1-[5-({[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}methyl)-4,5-dihydro-1,2-oxazol-3-yl]ethan-1-one

  • MDL number MFCD00202664
  • Cas Number : 338422-57-4
  • Molecular Weight : 321.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74802

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-945_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart